2,3,5-Trifluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,2,5-Trifluoro-3-iodobenzene

68868-1G 68868-25G 68868-5G 68868-500MG
76893-180EA 111.4 USD
76893-180 76893-182 76893-184 77651-932
2,3,5-Trifluoroiodobenzene ≥97%
2,3,5-Trifluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂F₃I
Storage Temperature: Refrigerator
MDL Number: MFCD08532463
CAS Number: 622379-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR