2,2,2-Trifluoro-4'-hydroxyacetophenone ≥97%
Supplier: AOBChem USA
|
Warning
|
67938-1G
67938-25G
67938-500MG
67938-5G
76893-152EA
137.92
USD
76893-152
76893-154
76893-156
76893-158
2,2,2-Trifluoro-4'-hydroxyacetophenone ≥97%
2,2,2-Trifluoro-4'-hydroxyacetophenone
Formula:
C₈H₅F₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD09832168 CAS Number: 1823-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...