1-(3,5-Bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

67111-1G 67111-25G 67111-5G
76893-128EA 112.46 USD
76893-128 76893-130 76893-132
1-(3,5-Bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,5-Bis(trifluoromethyl)phenyl)-2,2,2-trifluoroethanone
Formula: C₁₀H₃F₉O
MW: 310.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22688445
CAS Number: 130336-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR