1-Benzyl-2,4,5-tribromo-1H-imidazole ≥97%

Supplier: AOBChem USA

Warning

66458-25G 66458-5G
76893-106EA 460.44 USD
76893-106 76893-108
1-Benzyl-2,4,5-tribromo-1H-imidazole ≥97%
1-Benzyl-2,4,5-tribromo-1H-imidazole

Precaución: for Research Use Only

Formula: C₁₀H₇Br₃N₂
MW: 394.888 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411958
CAS Number: 31250-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR