2,4,5-Tribromo-1-(1-methylethyl)-1H-imidazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4,5-Tribromo-1-isopropyl-1H-imidazole

66454-1G 66454-25G 66454-500MG 66454-5G
76893-090EA 125.19 USD
76893-090 76893-092 76893-094 76893-096
2,4,5-Tribromo-1-(1-methylethyl)-1H-imidazole ≥97%
2,4,5-Tribromo-1-(1-methylethyl)-1H-imidazole

Precaución: for Research Use Only

Formula: C₆H₇Br₃N₂
MW: 346.845 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22690764
CAS Number: 863485-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR