1-Bromo-2-(difluoromethyl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

66288-1G 66288-25G 66288-500MG 66288-5G
76893-076EA 254.62 USD
76893-076 76893-078 76893-080 76893-082
1-Bromo-2-(difluoromethyl)-4-methylbenzene ≥97%
1-Bromo-2-(difluoromethyl)-4-methylbenzene
Formula: C₈H₇BrF₂
MW: 221.042 g/mol
Storage Temperature: Ambient
CAS Number: 1261442-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR