1-Chloro-4-(difluoromethyl)-2-fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

66262-1G 66262-25G 66262-5G
76893-064EA 77.45 USD
76893-064 76893-066 76893-068
1-Chloro-4-(difluoromethyl)-2-fluorobenzene ≥95%
1-Chloro-4-(difluoromethyl)-2-fluorobenzene
Formula: C₇H₄ClF₃
Storage Temperature: Ambient
MDL Number: MFCD14698548
CAS Number: 1214333-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR