1-(4-Ethylphenyl)-2,2,2-trifluoroethanol ≥97%

Supplier: AOBChem USA

Warning

65809-1G 65809-25G 65809-500MG 65809-5G
76893-048EA 219.61 USD
76893-048 76893-050 76893-052 76893-054
1-(4-Ethylphenyl)-2,2,2-trifluoroethanol ≥97%
1-(4-Ethylphenyl)-2,2,2-trifluoroethanol
Formula: C₁₀H₁₁F₃O
Storage Temperature: Ambient
CAS Number: 131252-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR