1-(2,6-Dimethyl-4-propoxyphenyl)-2,2,2-trifluoroethanone ≥97% research grade

Supplier: AOBChem USA

Warning

65715-1G 65715-25G 65715-500MG 65715-5G
76893-032EA 350.1 USD
76893-032 76893-034 76893-036 76893-038
1-(2,6-Dimethyl-4-propoxyphenyl)-2,2,2-trifluoroethanone ≥97% research grade
1-(2,6-Dimethyl-4-propoxyphenyl)-2,2,2-trifluoroethanone
Formula: C₁₃H₁₅F₃O₂
Storage Temperature: Refrigerator
CAS Number: 1443344-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR