2,2-Difluoro-1-(2-fluoropyridin-3-yl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65613-1G 65613-500MG 65613-5G
76893-022EA 967.55 USD
76893-022 76893-024 76893-026
2,2-Difluoro-1-(2-fluoropyridin-3-yl)ethanone ≥97%
2,2-Difluoro-1-(2-fluoropyridin-3-yl)ethanone
Formula: C₇H₄F₃NO
MW: 175.108 g/mol
Storage Temperature: Ambient
CAS Number: 2228138-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR