1-(3-Bromophenyl)-2,2-difluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
65587-1G
65587-25G
65587-500MG
65587-5G
76893-008EA
305.54
USD
76893-008
76893-010
76893-012
76893-014
1-(3-Bromophenyl)-2,2-difluoroethanone ≥97%
1-(3-Bromophenyl)-2,2-difluoroethanone
Formula:
C₈H₅BrF₂O MW: 235.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24447531 CAS Number: 1002356-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...