1-Bromo-5-chloro-4-iodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

65269-1G 65269-25G 65269-5G
76892-998EA 91.24 USD
76892-998 76893-000 76893-002
1-Bromo-5-chloro-4-iodo-2-methylbenzene ≥97%
1-Bromo-5-chloro-4-iodo-2-methylbenzene
Formula: C₇H₅BrClI
MW: 331.376 g/mol
Storage Temperature: Ambient
CAS Number: 1349718-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR