5'-Iodo-2',2,2,2-tetrafluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

65200-1G 65200-25G 65200-5G
76892-986EA 88.06 USD
76892-986 76892-988 76892-990
5'-Iodo-2',2,2,2-tetrafluoroacetophenone ≥97%
5'-Iodo-2',2,2,2-tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₃F₄IO
Storage Temperature: Ambient
MDL Number: MFCD08062422
CAS Number: 886502-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR