2,2,2-Trifluoro-1-(4-(methoxymethoxy)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

65117-1G 65117-25G 65117-500MG 65117-5G
76892-970EA 198.39 USD
76892-970 76892-972 76892-974 76892-976
2,2,2-Trifluoro-1-(4-(methoxymethoxy)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-(methoxymethoxy)phenyl)ethanone
Formula: C₁₀H₉F₃O₃
MW: 234.1718 g/mol
Storage Temperature: Refrigerator
CAS Number: 125680-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR