2,4-Dichloro-5-hydroxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

64934-1G 64934-25G 64934-500MG 64934-5G
76892-956EA 272.66 USD
76892-956 76892-958 76892-960 76892-962
2,4-Dichloro-5-hydroxybenzaldehyde ≥95%
2,4-Dichloro-5-hydroxybenzaldehyde
Formula: C₇H₄Cl₂O₂
MW: 191.0114 g/mol
Storage Temperature: Ambient
CAS Number: 56962-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR