2,4-Dichloro-6-fluoroanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1,5-Dichloro-3-fluoro-2-methoxybenzene

64419-1G 64419-25G 64419-500MG 64419-5G
76892-912EA 164.44 USD
76892-912 76892-914 76892-916 76892-918
2,4-Dichloro-6-fluoroanisole ≥97%
2,4-Dichloro-6-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO
MW: 195.018 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28785944
CAS Number: 54300-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR