1-Bromo-3-Chloro-5-(Methylsulfonyl)Benzene ≥97%

Supplier: AOBChem USA

Warning

64402-1G 64402-500MG 64402-5G
76892-898EA 525.15 USD
76892-898 76892-900 76892-902
1-Bromo-3-Chloro-5-(Methylsulfonyl)Benzene ≥97%
1-Bromo-3-Chloro-5-(Methylsulfonyl)Benzene
Formula: C₇H₆BrClO₂S
MW: 269.5433 g/mol
Storage Temperature: Ambient
CAS Number: 1779769-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR