1-(2-Fluoro-3-iodophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

64308-1G 64308-25G 64308-500MG 64308-5G
76892-884EA 173.99 USD
76892-884 76892-886 76892-888 76892-890
1-(2-Fluoro-3-iodophenyl)ethanone ≥97%
1-(2-Fluoro-3-iodophenyl)ethanone
Formula: C₈H₆FIO
MW: 264.0355 g/mol
Storage Temperature: Ambient
CAS Number: 1628003-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR