4'-Methoxy-2,2,2,3'-tetrafluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

64079-1G 64079-25G 64079-5G 64079-500MG
76892-860EA 115.64 USD
76892-860 76892-862 76892-864 77651-878
4'-Methoxy-2,2,2,3'-tetrafluoroacetophenone ≥97%
4'-Methoxy-2,2,2,3'-tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06201802
CAS Number: 132734-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR