4'-Chloro-2,2,2-trifluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

63831-25G 63831-5G
76892-852EA 289.63 USD
76892-852 76892-854
4'-Chloro-2,2,2-trifluoroacetophenone ≥97%
4'-Chloro-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD00018686
CAS Number: 321-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR