2',4'-Dimethyl-2,2,2-trifluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2,4-Dimethylphenyl)-2,2,2-trifluoroethanone

63751-1G 63751-25G 63751-5G
76892-842EA 85.94 USD
76892-842 76892-844 76892-846
2',4'-Dimethyl-2,2,2-trifluoroacetophenone ≥95%
2',4'-Dimethyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O
Storage Temperature: Ambient
MDL Number: MFCD01319984
CAS Number: 16184-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR