(2,6-Difluoro-3,5-dimethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

63707-1G 63707-25G 63707-5G
76892-824EA 117.76 USD
76892-824 76892-826 76892-828
(2,6-Difluoro-3,5-dimethoxyphenyl)methanol ≥97%
(2,6-Difluoro-3,5-dimethoxyphenyl)methanol
Formula: C₉H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD22683867
CAS Number: 1208434-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR