1-(Benzyloxy)-4-bromo-2-(tert-butyl)benzene ≥97%

Supplier: AOBChem USA

Warning

63662-1G 63662-25G 63662-500MG 63662-5G
76892-810EA 124.13 USD
76892-810 76892-812 76892-814 76892-816
1-(Benzyloxy)-4-bromo-2-(tert-butyl)benzene ≥97%
1-(Benzyloxy)-4-bromo-2-(tert-butyl)benzene
Formula: C₁₇H₁₉BrO
MW: 319.2361 g/mol
Storage Temperature: Ambient
CAS Number: 33839-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR