1-(2,3-Difluorophenyl)-2,2,2-trifluoroethanone ≥97%

Supplier: AOBChem USA

Warning

63452-1G 63452-25G 63452-500MG 63452-5G
76892-794EA 441.34 USD
76892-794 76892-796 76892-798 76892-800
1-(2,3-Difluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(2,3-Difluorophenyl)-2,2,2-trifluoroethanone
Formula: C₈H₃F₅O
MW: 210.1008 g/mol
Storage Temperature: Ambient
CAS Number: 1092712-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR