1,5-Dibromo-2-chloro-3-(trifluoromethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

63438-1G 63438-25G 63438-500MG 63438-5G
76892-778EA 206.88 USD
76892-778 76892-780 76892-782 76892-784
1,5-Dibromo-2-chloro-3-(trifluoromethoxy)benzene ≥95%
1,5-Dibromo-2-chloro-3-(trifluoromethoxy)benzene
Formula: C₇H₂Br₂ClF₃O
MW: 354.3463 g/mol
Storage Temperature: Ambient
CAS Number: 1000578-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR