2-(2-Aminothiazol-5-yl)-5-bromophenol ≥97%

Supplier: AOBChem USA

Warning

63371-1G 63371-25G 63371-500MG 63371-5G
76892-766EA 363.89 USD
76892-766 76892-768 76892-770 76892-772
2-(2-Aminothiazol-5-yl)-5-bromophenol ≥97%
2-(2-Aminothiazol-5-yl)-5-bromophenol
Formula: C₉H₇BrN₂OS
MW: 271.1336 g/mol
Storage Temperature: Ambient
CAS Number: 2807466-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR