4'-Methyl-2,2,2,3'-tetrafluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

63295-1G 63295-25G 63295-5G 63295-500MG
76892-748EA 99.73 USD
76892-748 76892-750 76892-752 77647-838
4'-Methyl-2,2,2,3'-tetrafluoroacetophenone ≥97%
4'-Methyl-2,2,2,3'-tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₆F₄O
Storage Temperature: Ambient
MDL Number: MFCD01320003
CAS Number: 845823-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR