2'-Bromo-3',6',2,2,2-pentafluoroacetophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2-Bromo-3,6-difluorophenyl)-2,2,2-trifluoroethanone

63283-1G 63283-500MG 63283-5G
76892-736EA 500.75 USD
76892-736 76892-738 76892-740
2'-Bromo-3',6',2,2,2-pentafluoroacetophenone ≥95%
2'-Bromo-3',6',2,2,2-pentafluoroacetophenone
Formula: C₈H₂BrF₅O
MW: 288.9968 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922615
CAS Number: 1208076-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR