1-Bromo-4-chloro-2-iodo-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

62046-1G 62046-25G 62046-5G
76892-554EA 66.84 USD
76892-554 76892-556 76892-558
1-Bromo-4-chloro-2-iodo-5-methylbenzene ≥97%
1-Bromo-4-chloro-2-iodo-5-methylbenzene
Formula: C₇H₅BrClI
MW: 331.376 g/mol
Storage Temperature: Ambient
CAS Number: 1255574-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR