1-Chloro-3-fluoro-4-(methoxymethoxy)-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

61969-1G 61969-25G 61969-500MG 61969-5G
76892-540EA 269.47 USD
76892-540 76892-542 76892-544 76892-546
1-Chloro-3-fluoro-4-(methoxymethoxy)-2-methylbenzene ≥97%
1-Chloro-3-fluoro-4-(methoxymethoxy)-2-methylbenzene
Formula: C₉H₁₀ClFO₂
MW: 204.6259 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32707259
CAS Number: 2432848-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR