(3-Methoxy-[1,1'-biphenyl]-4-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

61909-1G 61909-25G 61909-500MG 61909-5G
76892-524EA 381.93 USD
76892-524 76892-526 76892-528 76892-530
(3-Methoxy-[1,1'-biphenyl]-4-yl)boronic acid ≥97%
(3-Methoxy-[1,1'-biphenyl]-4-yl)boronic acid
Formula: C₁₃H₁₃BO₃
MW: 228.0515 g/mol
Storage Temperature: Ambient
CAS Number: 1379680-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR