1-Bromo-2-fluoro-5-nitro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

61403-1G 61403-25G 61403-500MG 61403-5G
76892-488EA 250.38 USD
76892-488 76892-490 76892-492 76892-494
1-Bromo-2-fluoro-5-nitro-3-(trifluoromethyl)benzene ≥97%
1-Bromo-2-fluoro-5-nitro-3-(trifluoromethyl)benzene
Formula: C₇H₂BrF₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD11520113
CAS Number: 1360438-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR