1,7-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

61359-1G 61359-25G 61359-500MG 61359-5G
76892-472EA 405.27 USD
76892-472 76892-474 76892-476 76892-478
1,7-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole ≥97%
1,7-Dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
Formula: C₁₅H₂₁BN₂O₂
MW: 272.1504 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30802377
CAS Number: 1572397-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR