1-Bromo-2-chloro-3,4,5-trifluorobenzene ≥95%

Supplier: AOBChem USA

Warning

61061-1G 61061-25G 61061-500MG 61061-5G
76892-444EA 149.59 USD
76892-444 76892-446 76892-448 76892-450
1-Bromo-2-chloro-3,4,5-trifluorobenzene ≥95%
1-Bromo-2-chloro-3,4,5-trifluorobenzene
Formula: C₆HBrClF₃
MW: 245.424 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070736
CAS Number: 122375-83-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR