4-(2-Chloroisonicotinoyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloropyridin-4-yl)(morpholino)methanone

60962-25G 60962-5G 60962-1G 60962-500MG
76892-428EA 516.66 USD
76892-428 76892-430 77639-826 77651-820
4-(2-Chloroisonicotinoyl)morpholine ≥97%
4-(2-Chloroisonicotinoyl)morpholine
Formula: C₁₀H₁₁ClN₂O₂
MW: 226.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864579
CAS Number: 174482-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR