2,3,6-Tribromopyridine ≥97%

Supplier: AOBChem USA

Warning

60787-25G 60787-5G
76892-420EA 410.57 USD
76892-420 76892-422
2,3,6-Tribromopyridine ≥97%
2,3,6-Tribromopyridine
Formula: C₅H₂Br₃N
Storage Temperature: Ambient
MDL Number: MFCD00234000
CAS Number: 2402-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR