2-Propyl-2H-pyrazole-3-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
1-Propyl-1H-pyrazole-5-carboxylic acid
60728-1G
60728-25G
60728-500MG
60728-5G
76892-404EA
123.07
USD
76892-404
76892-406
76892-408
76892-410
2-Propyl-2H-pyrazole-3-carboxylic acid ≥97%
2-Propyl-2H-pyrazole-3-carboxylic acid
Formula:
C₇H₁₀N₂O₂ MW: 154.17 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD04970195 CAS Number: 1004643-68-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...