1-(2-Methyl-4-(trifluoromethyl)phenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

60710-1G 60710-25G 60710-500MG 60710-5G
76892-392EA 321.46 USD
76892-392 76892-394 76892-396 76892-398
1-(2-Methyl-4-(trifluoromethyl)phenyl)ethanol ≥97%
1-(2-Methyl-4-(trifluoromethyl)phenyl)ethanol
Formula: C₁₀H₁₁F₃O
MW: 204.189 g/mol
Storage Temperature: Ambient
CAS Number: 1603075-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR