2-(4-Chloro-3-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

60453-1G 60453-25G 60453-500MG 60453-5G
76892-356EA 263.11 USD
76892-356 76892-358 76892-360 76892-362
2-(4-Chloro-3-methylphenyl)propan-2-ol ≥97%
2-(4-Chloro-3-methylphenyl)propan-2-ol
Formula: C₁₀H₁₃ClO
MW: 184.663 g/mol
Storage Temperature: Refrigerator
CAS Number: 1233333-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR