1-(4-Chloro-2-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

60381-1G 60381-25G 60381-5G
76892-326EA 111.4 USD
76892-326 76892-328 76892-330
1-(4-Chloro-2-methoxyphenyl)ethanone ≥95%
1-(4-Chloro-2-methoxyphenyl)ethanone
Formula: C₉H₉ClO₂
MW: 184.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11553429
CAS Number: 60207-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR