2-(Benzyloxy)-1-bromo-3-(tert-butyl)benzene ≥95%

Supplier: AOBChem USA

Warning

60202-1G 60202-25G 60202-500MG 60202-5G
76892-292EA 428.61 USD
76892-292 76892-294 76892-296 76892-298
2-(Benzyloxy)-1-bromo-3-(tert-butyl)benzene ≥95%
2-(Benzyloxy)-1-bromo-3-(tert-butyl)benzene
Formula: C₁₇H₁₉BrO
MW: 319.236 g/mol
Storage Temperature: Ambient
CAS Number: 2918924-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR