2,4-Dibromo-3-fluoro-6-methylaniline ≥97%

Supplier: AOBChem USA

Warning

60169-25G 60169-5G
76892-280EA 524.09 USD
76892-280 76892-282
2,4-Dibromo-3-fluoro-6-methylaniline ≥97%
2,4-Dibromo-3-fluoro-6-methylaniline
Formula: C₇H₆Br₂FN
MW: 282.936 g/mol
Storage Temperature: Ambient
CAS Number: 1349719-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR