2-(Cyclopropyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Cyclopropylphenyl)methanol

60159-1G 60159-25G 60159-500MG 60159-5G
76892-266EA 183.54 USD
76892-266 76892-268 76892-270 76892-272
2-(Cyclopropyl)benzyl alcohol ≥97%
2-(Cyclopropyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
Storage Temperature: Ambient
MDL Number: MFCD20921818
CAS Number: 118184-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR