2,4,6-Tribromopyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

60060-1G 60060-25G 60060-5G
76892-254EA 114.58 USD
76892-254 76892-256 76892-258
2,4,6-Tribromopyridin-3-ol ≥97%
2,4,6-Tribromopyridin-3-ol
Formula: C₅H₂Br₃NO
Storage Temperature: Refrigerator
CAS Number: 6602-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR