1-Bromo-2,5-dichloro-4-fluorobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

59344-1G 59344-25G 59344-500MG 59344-5G
76892-222EA 165.51 USD
76892-222 76892-224 76892-226 76892-228
1-Bromo-2,5-dichloro-4-fluorobenzene ≥97% research grade
1-Bromo-2,5-dichloro-4-fluorobenzene
Formula: C₆H₂BrCl₂F
MW: 243.89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845994
CAS Number: 1160573-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR