1-Bromo-3-fluoro-2-(methoxymethoxy)-5-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

58941-1G 58941-500MG 58941-5G
76892-186EA 521.97 USD
76892-186 76892-188 76892-190
1-Bromo-3-fluoro-2-(methoxymethoxy)-5-(trifluoromethyl)benzene ≥97%
1-Bromo-3-fluoro-2-(methoxymethoxy)-5-(trifluoromethyl)benzene
Formula: C₉H₇BrF₄O₂
MW: 303.048 g/mol
Storage Temperature: Refrigerator
CAS Number: 2764732-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR