1-Chloro-4-iodo-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

58475-1G 58475-25G 58475-5G 58475-500MG
76892-140EA 106.09 USD
76892-140 76892-142 76892-144 77647-766
1-Chloro-4-iodo-2,5-dimethylbenzene ≥97%
1-Chloro-4-iodo-2,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈ClI
Storage Temperature: Ambient
MDL Number: MFCD09955026
CAS Number: 854861-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR