3'-Methoxy-5'-(trifluoromethyl)acetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-(3-Methoxy-5-(trifluoromethyl)phenyl)ethanone

57951-1G 57951-25G 57951-5G 57951-500MG
76892-112EA 105.03 USD
76892-112 76892-114 76892-116 77651-766
3'-Methoxy-5'-(trifluoromethyl)acetophenone ≥97%
3'-Methoxy-5'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09025417
CAS Number: 916421-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR