2-(tert-Butyl)-4-fluorophenol ≥97%

Supplier: AOBChem USA

Warning

57706-1G 57706-25G 57706-500MG 57706-5G
76892-084EA 259.93 USD
76892-084 76892-086 76892-088 76892-090
2-(tert-Butyl)-4-fluorophenol ≥97%
2-(tert-Butyl)-4-fluorophenol
Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16657844
CAS Number: 357611-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR