2-Bromo-5-iodophenyl methyl sulfone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-Bromo-4-iodo-2-(methylsulphonyl)benzene

57691-1G 57691-25G 57691-500MG 57691-5G
76892-068EA 286.45 USD
76892-068 76892-070 76892-072 76892-074
2-Bromo-5-iodophenyl methyl sulfone ≥97%
2-Bromo-5-iodophenyl methyl sulfone

Precaución: for Research Use Only

Formula: C₇H₆BrIO₂S
MW: 360.995 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28155004
CAS Number: 1820619-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR